missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ELMO3 Partial ORF (AAH34410.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079767-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190057

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is similar to a C. elegans protein that functions in phagocytosis of apoptotic cells and in cell migration. Other members of this small family of engulfment and cell motility (ELMO) proteins have been shown to interact with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. [provided by RefSeq]

Sequence: MIFAREVISRNGLQILGTIIEDGDDLGEVLALSLRAFSELMEHGVVSWETLSIPFVRKVVCYVNMNLMDASVPPLALGLLESVTLSSPALGQLVKSEVPL
Specifications

Specifications

AAH34410.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MIFAREVISRNGLQILGTIIEDGDDLGEVLALSLRAFSELMEHGVVSWETLSIPFVRKVVCYVNMNLMDASVPPLALGLLESVTLSSPALGQLVKSEVPL
RUO
ELMO3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79767
ELMO3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CED-12/CED12/ELMO-3/FLJ13824
ELMO3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers