Learn More
Abnova™ Human ELMO1 Partial ORF (AAH03051, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009844-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. Similarity to a C. elegans protein suggests that this protein may function in apoptosis and in cell migration. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MQVVKEQVMRALTTKPSSLDQFKSKLQNLSYTEILKIRQSERMNQEDFQSRPILELKEKIQPEILELIKQQRLNRLVEGTCFRKLNARRRQDKFWYCRLSSpecifications
AAH03051 | |
Liquid | |
9844 | |
ELMO1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CED-12/CED12/ELMO-1/KIAA0281/MGC126406 | |
ELMO1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQVVKEQVMRALTTKPSSLDQFKSKLQNLSYTEILKIRQSERMNQEDFQSRPILELKEKIQPEILELIKQQRLNRLVEGTCFRKLNARRRQDKFWYCRLS | |
RUO | |
ELMO1 | |
Wheat Germ (in vitro) | |
GST |