missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EGLN3 Partial ORF (AAH10992.2, 94 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00112399-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: EMPLGHIMRLDLEKIALEYIVPCLHEVGFCYLDNFLGEVVGDCVLERVKQLHCTGALRDGQLAGPRAGVSKRHLRGDQITWIGGNEEGCEAISFLLSLIDSpecifications
AAH10992.2 | |
Liquid | |
112399 | |
EGLN3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ21620/HIFPH3/MGC125998/MGC125999/PHD3 | |
EGLN3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EMPLGHIMRLDLEKIALEYIVPCLHEVGFCYLDNFLGEVVGDCVLERVKQLHCTGALRDGQLAGPRAGVSKRHLRGDQITWIGGNEEGCEAISFLLSLID | |
RUO | |
EGLN3 | |
Wheat Germ (in vitro) | |
GST |