Learn More
Abnova™ Human EFHD1 Full-length ORF (AAH04128.2, 1 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080303-P01.10ug
Additional Details : Weight : 0.00010kg
Description
EFHD1 is an EF-hand domain-containing protein that displays increased expression during neuronal differentiation (Tominaga and Tomooka, 2002 [PubMed 12270117]).[supplied by OMIM]
Sequence: MASEELACKLERRLRREEAEESGPQLAPLGAPAPEPKPEPEPPARAPTASADAELSAQLSRRLDINEGAARPRRCRVFNPYTEFPEFSRRLIKDLESMFKLYDAGRDGFIDLMELKLMMEKLGAPQTHLGLKSMIKEVDEDFDGKLSFREFLLIFHKAAAGELQEDSGLMALAKLSEIDVALEGVRGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNTSpecifications
AAH04128.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781H0842/FLJ13612/MST133/MSTP133/PP3051 | |
EFHD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80303 | |
EFHD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASEELACKLERRLRREEAEESGPQLAPLGAPAPEPKPEPEPPARAPTASADAELSAQLSRRLDINEGAARPRRCRVFNPYTEFPEFSRRLIKDLESMFKLYDAGRDGFIDLMELKLMMEKLGAPQTHLGLKSMIKEVDEDFDGKLSFREFLLIFHKAAAGELQEDSGLMALAKLSEIDVALEGVRGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNT | |
RUO | |
EFHD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |