missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EDN1 Partial ORF (AAH09720, 113 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001906-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: SQKDKKCWNFCQAGKELRAEDIMEKDWNNHKKGKDCSKLGKKCIYQQLVRGRKIRRSSEEHLRQTRSETMRNSVKSSFHDPKLKGNPSRERYVTHNRAHWSpecifications
AAH09720 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SQKDKKCWNFCQAGKELRAEDIMEKDWNNHKKGKDCSKLGKKCIYQQLVRGRKIRRSSEEHLRQTRSETMRNSVKSSFHDPKLKGNPSRERYVTHNRAHW | |
RUO | |
EDN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1906 | |
EDN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ET1/HDLCQ7 | |
EDN1 | |
Recombinant | |
wheat germ expression system |