missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUPD1 Full-length ORF (NP_001003892.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00338599-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140124

  • 4130.00 NOK / 10µg
Estimated Shipment: 29-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTSGEVKTSLKNAYSSAKRLSPKMEEEGEEEDYCTPGAFELERLFWKGSPQYTHVNEVWPKLYIGDEATALDRYRLQKAGFTHVLNAAHGRWNVDTGPDYYRDMDIQYHGVEADDLPTFDLSVFFYPAAAFIDRALSDDHSKILVHCVMGRSRSATLVLAYLMIHKDMTLVDAIQQVAKNRCVLPNRGFLKQLRELDKQLVQQRRRSQRQDGEEEDGREL
Specifications

Specifications

NP_001003892.1
Liquid
338599
DUPD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTSGEVKTSLKNAYSSAKRLSPKMEEEGEEEDYCTPGAFELERLFWKGSPQYTHVNEVWPKLYIGDEATALDRYRLQKAGFTHVLNAAHGRWNVDTGPDYYRDMDIQYHGVEADDLPTFDLSVFFYPAAAFIDRALSDDHSKILVHCVMGRSRSATLVLAYLMIHKDMTLVDAIQQVAKNRCVLPNRGFLKQLRELDKQLVQQRRRSQRQDGEEEDGREL
RUO
DUPD1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DUSP27/FMDSP
DUPD1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers