Learn More
Abnova™ Human DSN1 Full-length ORF (AAH58899.1, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079980-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a kinetochore protein that functions as part of the minichromosome instability-12 centromere complex. The encoded protein is required for proper kinetochore assembly and progression through the cell cycle. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MTSVTRSEIIDEKGPVMSKTHDHQLESSLSPVEVFAKTSASLEMNQGVSEERIHLGSSPKKGGNCDLSHQERLQSKSLHLSPQEQSASYQDRRQSWRRASMKETNRRKSLHPIHQGITELSRSISVDLAESKRLGCLLLSSFQFSIQKLEPFLRDTKGFSLESFRAKASSLSEELKHFADGLETDGTLQKCFEDSNGKASDFSLEASVAEMKEYITKFSLERQTWDQLLLHYQQEAKEILSRGSTEAKITEVKVEPMTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPAIHGSGSGSCQSpecifications
AAH58899.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf172/FLJ13346/MGC32987/MIS13/dJ469A13.2 | |
DSN1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
79980 | |
DSN1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSVTRSEIIDEKGPVMSKTHDHQLESSLSPVEVFAKTSASLEMNQGVSEERIHLGSSPKKGGNCDLSHQERLQSKSLHLSPQEQSASYQDRRQSWRRASMKETNRRKSLHPIHQGITELSRSISVDLAESKRLGCLLLSSFQFSIQKLEPFLRDTKGFSLESFRAKASSLSEELKHFADGLETDGTLQKCFEDSNGKASDFSLEASVAEMKEYITKFSLERQTWDQLLLHYQQEAKEILSRGSTEAKITEVKVEPMTYLGSSQNEVLNTKPDYQKILQNQSKVFDCMELVMDELQGSVKQLQAFMDESTQCFQKVSVQLGKRSMQQLDPSPARKLLKLQLQNPPAIHGSGSGSCQ | |
RUO | |
DSN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |