missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DNAJB8 Full-length ORF (NP_699161.1, 1 a.a. - 232 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00165721-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16129023

  • 4115.00 NOK / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNAJB8 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM]

Sequence: MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSK
Specifications

Specifications

NP_699161.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DJ6/MGC33884
DNAJB8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
165721
DNAJB8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MANYYEVLGVQASASPEDIKKAYRKLALRWHPDKNPDNKEEAEKKFKLVSEAYEVLSDSKKRSLYDRAGCDSWRAGGGASTPYHSPFDTGYTFRNPEDIFREFFGGLDPFSFEFWDSPFNSDRGGRGHGLRGAFSAGFGEFPAFMEAFSSFNMLGCSGGSHTTFSSTSFGGSSSGSSGFKSVMSSTEMINGHKVTTKRIVENGQERVEVEEDGQLKSVTVNGKEQLKWMDSK
RUO
DNAJB8
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers