missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DKFZP434L1717 Partial ORF (NP_115512, 59 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084076-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: YKQTDPEHPDNDRFILSRGHAAPILYAAWVEVGDISESDLLNLRKLHSDLERHPTPRLPFVDSpecifications
NP_115512 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.56kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YKQTDPEHPDNDRFILSRGHAAPILYAAWVEVGDISESDLLNLRKLHSDLERHPTPRLPFVD | |
RUO | |
TKTL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84076 | |
DKFZP434L1717 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434L1717/FLJ32975 | |
TKTL2 | |
Recombinant | |
wheat germ expression system |