missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DIRAS3 Partial ORF (NP_004666, 130 a.a. - 229 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009077-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189875

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the ras superfamily, and is expressed in normal ovarian and breast epithelial cells but not in ovarian and breast cancers. It is a maternally imprinted gene and its expression is associated with growth suppression. Thus, this gene appears to be a putative tumor suppressor gene whose function is abrogated in ovarian and breast cancers. [provided by RefSeq]

Sequence: FYELICKIKGNNLHKFPIVLVGNKSDDTHREVALNDGATCAMEWNCAFMEISAKTDVNVQELFHMLLNYKKKPTTGLQEPEKKSQMPNTTEKLLDKCIIM
Specifications

Specifications

NP_004666
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FYELICKIKGNNLHKFPIVLVGNKSDDTHREVALNDGATCAMEWNCAFMEISAKTDVNVQELFHMLLNYKKKPTTGLQEPEKKSQMPNTTEKLLDKCIIM
RUO
DIRAS3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9077
DIRAS3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARHI/NOEY2
DIRAS3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers