Learn More
Abnova™ Human DHX9 Partial ORF (NP_001348, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001660-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein with RNA helicase activity. It may participate in melting of DNA:RNA hybrids, such as those that occur during transcription, and may play a role in X-linked gene expression. It contains 2 copies of a double-stranded RNA-binding domain, a DEXH core domain and an RGG box. The RNA-binding domains and RGG box influence and regulate RNA helicase activity. [provided by RefSeq]
Sequence: MGDVKNFLYAWCGKRKMTPSYEIRAVGNKNRQKFMCEVQVEGYNYTGMGNSTNKKDAQSNAARDFVNYLVRINEIKSEEVPAFGVASPPPSpecifications
NP_001348 | |
Liquid | |
1660 | |
DHX9 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX9/LKP/NDHII/RHA | |
DHX9 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGDVKNFLYAWCGKRKMTPSYEIRAVGNKNRQKFMCEVQVEGYNYTGMGNSTNKKDAQSNAARDFVNYLVRINEIKSEEVPAFGVASPPP | |
RUO | |
DHX9 | |
Wheat Germ (in vitro) | |
GST |