Learn More
Abnova™ Human DHDH Full-length ORF (NP_055290.1, 1 a.a. - 334 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027294-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an enzyme that belongs to the family of dihydrodiol dehydrogenases, which exist in multiple forms in mammalian tissues and are involved in the metabolism of xenobiotics and sugars. These enzymes catalyze the NADP1-linked oxidation of transdihydrodiols of aromatic hydrocarbons to corresponding catechols. This enzyme is a dimeric dihydrodiol dehydrogenase, and it differs from monomeric dihydrodiol dehydrogenases in its high substrate specificity for trans-dihydrodiols of aromatic hydrocarbons in the oxidative direction. [provided by RefSeq]
Sequence: MALRWGIVSVGLISSDFTAVLQTLPRSEHQVVAVAARDLSRAKEFAQKHDIPKAYGSYEELAKDPSVEVAYIGTQHPQHKAAVMLCLAAGKAVLCEKPTGVNAAEVREMVAEARSRALFLMEAIWTRFFPASEALRSVLAQGTLGDLRVARAEFGKNLIHVPRAVDRAQAGGALLDIGIYCVQFTSMVFGGQKPEKISVVGRRHETGVDDTVTVLLQYPGEVHGSFTCSITVQLSNTASVSGTKGMVQLLNPCWCPTELVVKGEHKEFPLPPVPKDCNFDNGAGMSYEAKHVWECLRKGMKESPVIPLSESELLADILEEVRKAIGVTFPQDKRSpecifications
NP_055290.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUM2DD | |
DHDH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27294 | |
DHDH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALRWGIVSVGLISSDFTAVLQTLPRSEHQVVAVAARDLSRAKEFAQKHDIPKAYGSYEELAKDPSVEVAYIGTQHPQHKAAVMLCLAAGKAVLCEKPTGVNAAEVREMVAEARSRALFLMEAIWTRFFPASEALRSVLAQGTLGDLRVARAEFGKNLIHVPRAVDRAQAGGALLDIGIYCVQFTSMVFGGQKPEKISVVGRRHETGVDDTVTVLLQYPGEVHGSFTCSITVQLSNTASVSGTKGMVQLLNPCWCPTELVVKGEHKEFPLPPVPKDCNFDNGAGMSYEAKHVWECLRKGMKESPVIPLSESELLADILEEVRKAIGVTFPQDKR | |
RUO | |
DHDH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |