Learn More
Abnova™ Human DGKA Full-length ORF (AAH31870, 1 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001606-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified. [provided by RefSeq]
Sequence: MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTLKDDGQHMWRPKRFPRPVYCNLCESSIGLGKQGLSCNLCKYTVHDQCAMKALPCEVSTYAKSRKDIGVQSHVWVRGGCESGRCDRCQKKIRIYHSLTGLHCVWCHLEIHDDCLQAVGHECDCGLLRDHILPPSSIYPSVLASGPDRKNSKTSQKTMDDLNLSTSEALRIDPVPNTHPLLVFVNPKSGGKQGQRVLWKFQYILNPRQVFNLLKDGPEIGLRLFKDVPDSRILVCGGDGTVGWILETIDKANLPVLPPVAVLPLGTGNDLARCLRWGGGYEGQNLAKILKDLEMSKVVHMDRWSVEVIPQQTEEKSDPVPFQIINNYFSIGVDASIAHRFYIMREKYPEKFNSRMKNKLWYFEFATSESIFSTCKKLEESLTVEICGKPLDLSNLSLEGIAVLNIPSMHGGSNLWGDTRRPHGDIYGINQALGATAKVITDPDILKTCVPDLSDKRLEVVGLEGAIEMGQIYTKLKNAGRRLAKCSEITFHTTKTLPMQIDGEPWVQTPCTIKITHKNQMPMLMGPPPRSTNFFGFLSSpecifications
AAH31870 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.37kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAGK/DAGK1/DGK-alpha/MGC12821/MGC42356 | |
DGKA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1606 | |
DGKA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKERGLISPSDFAQLQKYMEYSTKKVSDVLKLFEDGEMAKYVQGDAIGYEGFQQFLKIYLEVDNVPRHLSLALFQSFETGHCLNETNVTKDVVCLNDVSCYFSLLEGGRPEDKLEFTFKLYDTDRNGILDSSEVDKIILQMMRVAEYLDWDVSELRPILQEMMKEIDYDGSGSVSQAEWVRAGATTVPLLVLLGLEMTLKDDGQHMWRPKRFPRPVYCNLCESSIGLGKQGLSCNLCKYTVHDQCAMKALPCEVSTYAKSRKDIGVQSHVWVRGGCESGRCDRCQKKIRIYHSLTGLHCVWCHLEIHDDCLQAVGHECDCGLLRDHILPPSSIYPSVLASGPDRKNSKTSQKTMDDLNLSTSEALRIDPVPNTHPLLVFVNPKSGGKQGQRVLWKFQYILNPRQVFNLLKDGPEIGLRLFKDVPDSRILVCGGDGTVGWILETIDKANLPVLPPVAVLPLGTGNDLARCLRWGGGYEGQNLAKILKDLEMSKVVHMDRWSVEVIPQQTEEKSDPVPFQIINNYFSIGVDASIAHRFYIMREKYPEKFNSRMKNKLWYFEFATSESIFSTCKKLEESLTVEICGKPLDLSNLSLEGIAVLNIPSMHGGSNLWGDTRRPHGDIYGINQALGATAKVITDPDILKTCVPDLSDKRLEVVGLEGAIEMGQIYTKLKNAGRRLAKCSEITFHTTKTLPMQIDGEPWVQTPCTIKITHKNQMPMLMGPPPRSTNFFGFLS | |
RUO | |
DGKA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |