Learn More
Abnova™ Human DGCR8 Partial ORF (NP_073557, 675 a.a. - 771 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00054487-Q01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16127586
  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
Description
Sequence: LASQKILQLLHPHVKNWGSLLRMYGRESSKMVKQETSDKSVIELQQYAKKNKPNLHILSKLQEEMKRLAEEREETRKKPKMSIVASAQPGGEPLCTV
Specifications
Specifications
NP_073557
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LASQKILQLLHPHVKNWGSLLRMYGRESSKMVKQETSDKSVIELQQYAKKNKPNLHILSKLQEEMKRLAEEREETRKKPKMSIVASAQPGGEPLCTV
RUO
DGCR8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
54487
DGCR8 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C22orf12/DGCRK6/Gy1
DGCR8
Recombinant
wheat germ expression system