Learn More
Abnova™ Human DDX56 Full-length ORF (NP_061955.1, 1 a.a. - 547 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054606-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The protein encoded by this gene shows ATPase activity in the presence of polynucleotides and associates with nucleoplasmic 65S preribosomal particles. This gene may be involved in ribosome synthesis, most likely during assembly of the large 60S ribosomal subunit. [provided by RefSeq]
Sequence: MEDSEALGFEHMGLDPRLLQAVTDLGWSRPTLIQEKAIPLALEGKDLLARARTGSGKTAAYAIPMLQLLLHRKATGPVVEQAVRGLVLVPTKELARQAQSMIQQLATYCARDVRVANVSAAEDSVSQRAVLMEKPDVVVGTPSRILSHLQQDSLKLRDSLELLVVDEADLLFSFGFEEELKSLLCHLPRIYQAFLMSATFNEDVQALKELILHNPVTLKLQESQLPGPDQLQQFQVVCETEEDKFLLLYALLKLSLIRGKSLLFVNTLERSYRLRLFLEQFSIPTCVLNGELPLRSRCHIISQFNQGFYDCVIATDAEVLGAPVKGKRRGRGPKGDKASDPEAGVARGIDFHHVSAVLNFDLPPTPEAYIHRAGRTARANNPGIVLTFVLPTEQFHLGKIEELLSGENRGPILLPYQFRMEEIEGFRYRCRDAMRSVTKQAIREARLKEIKEELLHSEKLKTYFEDNPRDLQLLRHDLPLHPAVVKPHLGHVPDYLVPPALRGLVRPHKKRKKLSSSCRKAKRAKSQNPLRSFKHKGKKFRPTAKPSSpecifications
NP_061955.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX21/DDX26/NOH61 | |
DDX56 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54606 | |
DDX56 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEDSEALGFEHMGLDPRLLQAVTDLGWSRPTLIQEKAIPLALEGKDLLARARTGSGKTAAYAIPMLQLLLHRKATGPVVEQAVRGLVLVPTKELARQAQSMIQQLATYCARDVRVANVSAAEDSVSQRAVLMEKPDVVVGTPSRILSHLQQDSLKLRDSLELLVVDEADLLFSFGFEEELKSLLCHLPRIYQAFLMSATFNEDVQALKELILHNPVTLKLQESQLPGPDQLQQFQVVCETEEDKFLLLYALLKLSLIRGKSLLFVNTLERSYRLRLFLEQFSIPTCVLNGELPLRSRCHIISQFNQGFYDCVIATDAEVLGAPVKGKRRGRGPKGDKASDPEAGVARGIDFHHVSAVLNFDLPPTPEAYIHRAGRTARANNPGIVLTFVLPTEQFHLGKIEELLSGENRGPILLPYQFRMEEIEGFRYRCRDAMRSVTKQAIREARLKEIKEELLHSEKLKTYFEDNPRDLQLLRHDLPLHPAVVKPHLGHVPDYLVPPALRGLVRPHKKRKKLSSSCRKAKRAKSQNPLRSFKHKGKKFRPTAKPS | |
RUO | |
DDX56 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |