missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDEF2 Partial ORF (NP_003878, 909 a.a. - 1006 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008853-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16189465

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a multidomain protein containing an N-terminal alpha-helical region with a coiled-coil motif, followed by a pleckstrin homology (PH) domain, an Arf-GAP domain, an ankyrin homology region, a proline-rich region, and a C-terminal Src homology 3 (SH3) domain. The protein localizes in the Golgi apparatus and at the plasma membrane, where it colocalizes with protein tyrosine kinase 2-beta (PYK2). The encoded protein forms a stable complex with PYK2 in vivo. This interaction appears to be mediated by binding of its SH3 domain to the C-terminal proline-rich domain of PYK2. The encoded protein is tyrosine phosphorylated by activated PYK2. It has catalytic activity for class I and II ArfGAPs in vitro, and can bind the class III Arf ARF6 without immediate GAP activity. The encoded protein is believed to function as an ARF GAP that controls ARF-mediated vesicle budding when recruited to Golgi membranes. In addition, it functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: RGPVDLSATEALGPLSNAMVLQPPAPMPRKSQATKLKPKRVKALYNCVADNPDELTFSEGDVIIVDGEEDQEWWIGHIDGDPGRKGAFPVSFVHFIAD
Specifications

Specifications

NP_003878
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RGPVDLSATEALGPLSNAMVLQPPAPMPRKSQATKLKPKRVKALYNCVADNPDELTFSEGDVIIVDGEEDQEWWIGHIDGDPGRKGAFPVSFVHFIAD
RUO
ASAP2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8853
DDEF2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AMAP2/CENTB3/DDEF2/FLJ42910/KIAA0400/PAG3/PAP/Pap-alpha/SHAG1
ASAP2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers