missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CYTH2 Partial ORF (NP_059431, 314 a.a. - 398 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009266-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140296

  • 4115.00 NOK / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: VDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQE
Specifications

Specifications

NP_059431
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.09kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VDDPRKPNCFELYIPNNKGQLIKACKTEADGRVVEGNHMVYRISAPTQEEKDEWIKSIQAAVSVDPFYEMLAARKKRISVKKKQE
RUO
CYTH2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9266
CYTH2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARNO/CTS18/CTS18.1/PSCD2/PSCD2L/SEC7L/Sec7p-L/Sec7p-like
CYTH2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers