Learn More
Abnova™ Human CYP26C1 Partial ORF (NP_899230.2, 423 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00340665-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is involved in the catabolism of all-trans- and 9-cis-retinoic acid, and thus contributes to the regulation of retinoic acid levels in cells and tissues. This gene is adjacent to a related gene on chromosome 10q23.33. [provided by RefSeq]
Sequence: SPPEGFDPERFGAAREDSRGASSRFHYIPFGGGARSCLGQELAQAVLQLLAVELVRTARWELATPAFPAMQTVPIVHPVDGLRLFFHPLTPSVAGNGLCLSpecifications
NP_899230.2 | |
Liquid | |
340665 | |
CYP26C1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ45301 | |
CYP26C1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SPPEGFDPERFGAAREDSRGASSRFHYIPFGGGARSCLGQELAQAVLQLLAVELVRTARWELATPAFPAMQTVPIVHPVDGLRLFFHPLTPSVAGNGLCL | |
RUO | |
CYP26C1 | |
Wheat Germ (in vitro) | |
GST |