missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CXCL12 (SDF-1a) Recombinant Protein

Product Code. p-7220362
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
This item is not returnable. View return policy

Product Code. 17656655

Brand: Invitrogen™ RP87861

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48061
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation 20 mM tris with 0.5 M NaCl and no preservative; pH 8
Gene ID (Entrez) 6387
Molecular Weight (g/mol) 8 kD
Name Human CXCL12 alpha (SDF-1 alpha) (aa 22-89)
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.1 EU/ μg cytokine
Gene Alias 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; PBSF; Pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 A; SDF 1 alpha; SDF 1 A; Sdf1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; TPAR1
Gene Symbol CXCL12
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.