Learn More
Abnova™ Human CUTL1 Partial ORF (NP_001904.2, 521 a.a. - 620 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001523-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: AENRLAQHTLQALQSELDSLRADNIKLFEKIKFLQSYPGRGSGSDDTELRYSSQYEERLDPFSSFSKRERQRKYLSLSPWDKATLSMGRLVLSNKMARTISpecifications
NP_001904.2 | |
Liquid | |
1523 | |
CUTL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CASP/CDP/CDP/Cut/CDP1/COY1/CUTL1/CUX/Clox/Cux/CDP/GOLIM6/Nbla10317/p100/p110/p200/p75 | |
CUX1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AENRLAQHTLQALQSELDSLRADNIKLFEKIKFLQSYPGRGSGSDDTELRYSSQYEERLDPFSSFSKRERQRKYLSLSPWDKATLSMGRLVLSNKMARTI | |
RUO | |
CUX1 | |
Wheat Germ (in vitro) | |
GST |