missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CTDSPL Full-length ORF (NP_005799.2, 1 a.a. - 265 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010217-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MDGPAIITQVTNPKEDEGRLPGAGEKASQCNVSLKKQRSRSILSSFFCCFRDYNVEAPPPSSPSVLPPLVEENGGLQKPPAKYLLPEVTVLDYGKKCVVIDLDETLVHSSFKPISNADFIVPVEIDGTIHQVYVLKRPHVDEFLQRMGQLFECVLFTASLAKYADPVADLLDRWGVFRARLFRESCVFHRGNYVKDLSRLGRELSKVIIVDNSPASYIFHPENAVPVQSWFDDMTDTELLDLIPFFEGLSREDDVYSMLHRLCNRSpecifications
NP_005799.2 | |
Liquid | |
10217 | |
CTDSPL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDGPAIITQVTNPKEDEGRLPGAGEKASQCNVSLKKQRSRSILSSFFCCFRDYNVEAPPPSSPSVLPPLVEENGGLQKPPAKYLLPEVTVLDYGKKCVVIDLDETLVHSSFKPISNADFIVPVEIDGTIHQVYVLKRPHVDEFLQRMGQLFECVLFTASLAKYADPVADLLDRWGVFRARLFRESCVFHRGNYVKDLSRLGRELSKVIIVDNSPASYIFHPENAVPVQSWFDDMTDTELLDLIPFFEGLSREDDVYSMLHRLCNR | |
RUO | |
CTDSPL | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
56.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3orf8/HYA22/PSR1/RBSP3/SCP3 | |
CTDSPL | |
Yes | |
wheat germ expression system |