Learn More
Abnova™ Human CRHBP Full-length ORF (NP_001873.2, 1 a.a. - 322 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001393-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Corticotropin-releasing hormone is a potent stimulator of synthesis and secretion of preopiomelanocortin-derived peptides. Although CRH concentrations in the human peripheral circulation are normally low, they increase throughout pregnancy and fall rapidly after parturition. Maternal plasma CRH probably originates from the placenta. Human plasma contains a CRH-binding protein which inactivates CRH and which may prevent inappropriate pituitary-adrenal stimulation in pregnancy. [provided by RefSeq]
Sequence: MSPNFKLQCHFILIFLTALRGESRYLELREAADYDPFLLFSANLKRELAGEQPYRRALRCLDMLSLQGQFTFTADRPQLHCAAFFISEPEEFITIHYDQVSIDCQGGDFLKVFDGWILKGEKFPSSQDHPLPSAERYIDFCESGLSRRSIRSSQNVAMIFFRVHEPGNGFTLTIKTDPNLFPCNVISQTPNGKFTLVVPHQHRNCSFSIIYPVVIKISDLTLGHVNGLQLKKSSAGCEGIGDFVELLGGTGLDPSKMTPLADLCYPFHGPAQMKVGCDNTVVRMVSSGKHVNRVTFEYRQLEPYELENPNGNSIGEFCLSGLSpecifications
NP_001873.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CRF-BP/CRFBP | |
CRHBP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1393 | |
CRHBP (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSPNFKLQCHFILIFLTALRGESRYLELREAADYDPFLLFSANLKRELAGEQPYRRALRCLDMLSLQGQFTFTADRPQLHCAAFFISEPEEFITIHYDQVSIDCQGGDFLKVFDGWILKGEKFPSSQDHPLPSAERYIDFCESGLSRRSIRSSQNVAMIFFRVHEPGNGFTLTIKTDPNLFPCNVISQTPNGKFTLVVPHQHRNCSFSIIYPVVIKISDLTLGHVNGLQLKKSSAGCEGIGDFVELLGGTGLDPSKMTPLADLCYPFHGPAQMKVGCDNTVVRMVSSGKHVNRVTFEYRQLEPYELENPNGNSIGEFCLSGL | |
RUO | |
CRHBP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |