Learn More
Abnova™ Human COX4I2 Full-length ORF (AAH57779, 1 a.a. - 171 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084701-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes isoform 2 of subunit IV. Isoform 1 of subunit IV is encoded by a different gene, however, the two genes show a similar structural organization. Subunit IV is the largest nuclear encoded subunit which plays a pivotal role in COX regulation. [provided by RefSeq]
Sequence: MLPRAAWSLVLRKGGGGRRGMHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKTVMGCVFFFIGFAALVIWWQRVYVFPPKPITLTDERKAQQLQRMLDMKVNPVQGLASRWDYEKKQWKKSpecifications
AAH57779 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.55kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COX4/COX4-2/COX4B/COX4L2/COXIV-2/dJ857M17.2 | |
COX4I2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84701 | |
COX4I2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLPRAAWSLVLRKGGGGRRGMHSSEGTTRGGGKMSPYTNCYAQRYYPMPEEPFCTELNAEEQALKEKEKGSWTQLTHAEKVALYRLQFNETFAEMNRRSNEWKTVMGCVFFFIGFAALVIWWQRVYVFPPKPITLTDERKAQQLQRMLDMKVNPVQGLASRWDYEKKQWKK | |
RUO | |
COX4I2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |