missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COUP-TF II/NR2F2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems PP-H7147-00
This item is not returnable.
View return policy
Description
COUP-TF II/NR2F2 Monoclonal specifically detects COUP-TF II/NR2F2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| COUP-TF II/NR2F2 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| NR2F2 | |
| Recombinant human COUP-TF II/NR2F2 Amino Acids 43-64 | |
| 100 μg | |
| Primary | |
| This antibody specifically recognizes human COUP-TF II and cross-reacts with rat COUP-TF II. This antibody does not cross-react with human COUP-TF I or EAR2. Not yet tested in other species. | |
| This antibody is stable for greater than six months when held at -20degrees C in a manual defrost freezer or at -70degrees C. Upon thawing, the antibody can be stored at 2degrees - 8degrees C for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| H7147 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Apolipoprotein A-I regulatory protein 1, ARP-1, ARP1ADP-ribosylation factor related protein 1, chicken ovalbumin upstream promoter transcription factor 2, COUP transcription factor 2, COUP transcription factor II, COUPTF II, COUP-TF II, COUP-TF2, COUPTFB, COUPTFII, COUP-TFII, MGC117452, NR2F2, Nuclear receptor subfamily 2 group F member 2, nuclear receptor subfamily 2, group F, member 2, SVP40, TFCOUP2apolipoprotein AI regulatory protein 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7026 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction