missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (279) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14662JF635
This item is not returnable.
View return policy
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-NL63 samples. It is validated for ELISA
Specifications
| Human Coronavirus Spike Protein | |
| Monoclonal | |
| Janelia Fluor 635 | |
| HKU1, Spike glycoprotein | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| HCoV-NL63 | |
| Purified |
| ELISA | |
| 279 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3200426 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction