Learn More
Abnova™ Human COPB2 Partial ORF (NP_004757.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009276-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The Golgi coatomer complex (see MIM 601924) constitutes the coat of nonclathrin-coated vesicles and is essential for Golgi budding and vesicular trafficking. It consists of 7 protein subunits, including COPB2.[supplied by OMIM]
Sequence: MPLRLDIKRKLTARSDRVKSVDLHPTEPWMLASLYNGSVCVWNHETQTLVKTFEVCDLPVRAAKFVARKNWVVTGADDMQIRVFNYNTLERVHMFEAHSSpecifications
NP_004757.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPLRLDIKRKLTARSDRVKSVDLHPTEPWMLASLYNGSVCVWNHETQTLVKTFEVCDLPVRAAKFVARKNWVVTGADDMQIRVFNYNTLERVHMFEAHS | |
RUO | |
COPB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9276 | |
COPB2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
beta'-COP | |
COPB2 | |
Recombinant | |
wheat germ expression system |