Learn More
Abnova™ Human COLEC10 Partial ORF (NP_006429.1, 121 a.a. - 229 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010584-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. The other members of this family are secreted proteins and bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. This gene product is a cytosolic protein, a characteristic that suggests that it may have different biological functions than other C-lectins. [provided by RefSeq]
Sequence: CGRYRKFVGQLDISIARLKTSMKFVKNVIAGIRETEEKFYYIVQEEKNYRESLTHCRIRGGMLAMPKDEAANTLIADYVAKSGFFRVFIGVNDLEREGQYMFTDNTPLQSpecifications
NP_006429.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CGRYRKFVGQLDISIARLKTSMKFVKNVIAGIRETEEKFYYIVQEEKNYRESLTHCRIRGGMLAMPKDEAANTLIADYVAKSGFFRVFIGVNDLEREGQYMFTDNTPLQ | |
RUO | |
COLEC10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10584 | |
COLEC10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLL1/MGC118794/MGC118795 | |
COLEC10 | |
Recombinant | |
wheat germ expression system |