Learn More
Abnova™ Human COG3 Full-length ORF (NP_113619.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083548-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles. [provided by RefSeq]
Sequence: MAEAALLLLPEAAAERDAREKLALWDRRPDTTAPLTDRQTDSVLELKAAAENLPVPAELPIEDLCSLTSQSLPIELTSVVPESTEDILLKGFTSLGMEEERIETAQQFFSWFAKLQTQMDQDEGTKYRQMRDYLSGFQEQCDAILNDVNSALQHLESLQKQYLFVSNKTGTLHEACEQLLKEQSELVDLAENIQQKLSYFNELETINTKLNSPTLSVNSDGFIPMLAKLDDCITYISSHPNFKDYPIYLLKFKQCLSKALHLMKTYTVNTLQTLTSQLLKRDPSSVPNADNAFTLFYVKFRAAAPKVRTLIEQIELRSEKIPEYQQLLNDIHQCYLDQRELLLGPSIACTVAELTSQNNRDHCALVRSGCAFMVHVCQDEHQLYNEFFTKPTSKLDELLEKLCVSLYDVFRPLIIHVIHLETLSELCGILKNEVLEDHVQNNAEQLGAFAAGVKQMLEDVQERLVYRTHIYIQTDITGYKPAPGDLAYPDKLVMMEQIAQSLKDEQKKVPSEASFSDVHLEEGESNSLTKSGSTESLNPRPQTTISPADLHGMWYPTVRRTLVCLSKLYRCIDRAVFQGLSQEALSACIQSLLGASESISKNKTQIDGQLFLIKHLLILREQIAPFHTEFTIKEISLDLKKTRDAAFKILNPMTVPRFFRLNSNNALIEFLLEGTPEIREHYLDSKKDVDRHLKSACEQFIQQQTKLFVEQLEEFMTKVSALKTMASQGGPKYTLSQQPWAQPAKVSDLAATAYKTIKTKLPVTLRSMSLYLSNKDTEFILFKPVRNNIQQVFQKFHALLKEEFSPEDIQIIACPSMEQLSLLLSVSKSpecifications
NP_113619.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
120.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEC34 | |
COG3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83548 | |
COG3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEAALLLLPEAAAERDAREKLALWDRRPDTTAPLTDRQTDSVLELKAAAENLPVPAELPIEDLCSLTSQSLPIELTSVVPESTEDILLKGFTSLGMEEERIETAQQFFSWFAKLQTQMDQDEGTKYRQMRDYLSGFQEQCDAILNDVNSALQHLESLQKQYLFVSNKTGTLHEACEQLLKEQSELVDLAENIQQKLSYFNELETINTKLNSPTLSVNSDGFIPMLAKLDDCITYISSHPNFKDYPIYLLKFKQCLSKALHLMKTYTVNTLQTLTSQLLKRDPSSVPNADNAFTLFYVKFRAAAPKVRTLIEQIELRSEKIPEYQQLLNDIHQCYLDQRELLLGPSIACTVAELTSQNNRDHCALVRSGCAFMVHVCQDEHQLYNEFFTKPTSKLDELLEKLCVSLYDVFRPLIIHVIHLETLSELCGILKNEVLEDHVQNNAEQLGAFAAGVKQMLEDVQERLVYRTHIYIQTDITGYKPAPGDLAYPDKLVMMEQIAQSLKDEQKKVPSEASFSDVHLEEGESNSLTKSGSTESLNPRPQTTISPADLHGMWYPTVRRTLVCLSKLYRCIDRAVFQGLSQEALSACIQSLLGASESISKNKTQIDGQLFLIKHLLILREQIAPFHTEFTIKEISLDLKKTRDAAFKILNPMTVPRFFRLNSNNALIEFLLEGTPEIREHYLDSKKDVDRHLKSACEQFIQQQTKLFVEQLEEFMTKVSALKTMASQGGPKYTLSQQPWAQPAKVSDLAATAYKTIKTKLPVTLRSMSLYLSNKDTEFILFKPVRNNIQQVFQKFHALLKEEFSPEDIQIIACPSMEQLSLLLSVSK | |
RUO | |
COG3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |