missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COG3 Full-length ORF (NP_113619.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00083548-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135373

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene has similarity to a yeast protein. It seems to be part of a peripheral membrane protein complex localized on cis/medial Golgi cisternae where it may participate in tethering intra-Golgi transport vesicles. [provided by RefSeq]

Sequence: MAEAALLLLPEAAAERDAREKLALWDRRPDTTAPLTDRQTDSVLELKAAAENLPVPAELPIEDLCSLTSQSLPIELTSVVPESTEDILLKGFTSLGMEEERIETAQQFFSWFAKLQTQMDQDEGTKYRQMRDYLSGFQEQCDAILNDVNSALQHLESLQKQYLFVSNKTGTLHEACEQLLKEQSELVDLAENIQQKLSYFNELETINTKLNSPTLSVNSDGFIPMLAKLDDCITYISSHPNFKDYPIYLLKFKQCLSKALHLMKTYTVNTLQTLTSQLLKRDPSSVPNADNAFTLFYVKFRAAAPKVRTLIEQIELRSEKIPEYQQLLNDIHQCYLDQRELLLGPSIACTVAELTSQNNRDHCALVRSGCAFMVHVCQDEHQLYNEFFTKPTSKLDELLEKLCVSLYDVFRPLIIHVIHLETLSELCGILKNEVLEDHVQNNAEQLGAFAAGVKQMLEDVQERLVYRTHIYIQTDITGYKPAPGDLAYPDKLVMMEQIAQSLKDEQKKVPSEASFSDVHLEEGESNSLTKSGSTESLNPRPQTTISPADLHGMWYPTVRRTLVCLSKLYRCIDRAVFQGLSQEALSACIQSLLGASESISKNKTQIDGQLFLIKHLLILREQIAPFHTEFTIKEISLDLKKTRDAAFKILNPMTVPRFFRLNSNNALIEFLLEGTPEIREHYLDSKKDVDRHLKSACEQFIQQQTKLFVEQLEEFMTKVSALKTMASQGGPKYTLSQQPWAQPAKVSDLAATAYKTIKTKLPVTLRSMSLYLSNKDTEFILFKPVRNNIQQVFQKFHALLKEEFSPEDIQIIACPSMEQLSLLLSVSK
Specifications

Specifications

NP_113619.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
120.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SEC34
COG3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
83548
COG3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEAALLLLPEAAAERDAREKLALWDRRPDTTAPLTDRQTDSVLELKAAAENLPVPAELPIEDLCSLTSQSLPIELTSVVPESTEDILLKGFTSLGMEEERIETAQQFFSWFAKLQTQMDQDEGTKYRQMRDYLSGFQEQCDAILNDVNSALQHLESLQKQYLFVSNKTGTLHEACEQLLKEQSELVDLAENIQQKLSYFNELETINTKLNSPTLSVNSDGFIPMLAKLDDCITYISSHPNFKDYPIYLLKFKQCLSKALHLMKTYTVNTLQTLTSQLLKRDPSSVPNADNAFTLFYVKFRAAAPKVRTLIEQIELRSEKIPEYQQLLNDIHQCYLDQRELLLGPSIACTVAELTSQNNRDHCALVRSGCAFMVHVCQDEHQLYNEFFTKPTSKLDELLEKLCVSLYDVFRPLIIHVIHLETLSELCGILKNEVLEDHVQNNAEQLGAFAAGVKQMLEDVQERLVYRTHIYIQTDITGYKPAPGDLAYPDKLVMMEQIAQSLKDEQKKVPSEASFSDVHLEEGESNSLTKSGSTESLNPRPQTTISPADLHGMWYPTVRRTLVCLSKLYRCIDRAVFQGLSQEALSACIQSLLGASESISKNKTQIDGQLFLIKHLLILREQIAPFHTEFTIKEISLDLKKTRDAAFKILNPMTVPRFFRLNSNNALIEFLLEGTPEIREHYLDSKKDVDRHLKSACEQFIQQQTKLFVEQLEEFMTKVSALKTMASQGGPKYTLSQQPWAQPAKVSDLAATAYKTIKTKLPVTLRSMSLYLSNKDTEFILFKPVRNNIQQVFQKFHALLKEEFSPEDIQIIACPSMEQLSLLLSVSK
RUO
COG3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers