Learn More
Abnova™ Human COG1 Partial ORF (NP_061184.1, 881 a.a. - 980 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009382-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is one of eight proteins (Cog1-8) which form a Golgi-localized complex (COG) required for normal Golgi morphology and function. It is thought that this protein is required for steps in the normal medial and trans Golgi-associated processing of glycoconjugates and plays a role in the organization of the Golgi-localized complex. [provided by RefSeq]
Sequence: GTENQLAPRSSTFNSQEPHNILPLASSQIRFGLLPLSMTSTRKAKSTRNIETKAQVVPPARSTAGDPTVPGSLFRQLVSEEDNTSAPSLFKLGWLSSMTKSpecifications
NP_061184.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTENQLAPRSSTFNSQEPHNILPLASSQIRFGLLPLSMTSTRKAKSTRNIETKAQVVPPARSTAGDPTVPGSLFRQLVSEEDNTSAPSLFKLGWLSSMTK | |
RUO | |
COG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9382 | |
COG1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDG2G/DKFZp762L1710/KIAA1381/LDLB | |
COG1 | |
Recombinant | |
wheat germ expression system |