missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor III/Tissue Factor Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2339
This item is not returnable.
View return policy
Description
Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| Coagulation Factor III/Tissue Factor | |
| Polyclonal | |
| Biotin | |
| P13726 | |
| F3 | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III/Tissue Factor (R&D Systems, Catalog # 2339-PA) Gly34-Glu251 Accession # P13726 | |
| 50 μg | |
| Primary | |
| Detects human Coagulation Factor III/Tissue Factor in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with human Plasminogen, recombinant human (rh) Coagulation Factor VII, rhCoagulation Factor X, rhCoagulation Factor XI, rhCoagulation Factor XIV and recombinant mouse Coagulation Factor III/Tissue Factor is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2152 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction