missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CNTNAP5 Partial ORF (NP_570129, 1062 a.a. - 1160 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00129684-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16151767

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene product belongs to the neurexin family, members of which function in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, and thrombospondin N-terminal-like domains. [provided by RefSeq]

Sequence: DFVVVLLCKNGSLQVRYHLNKEETHVFTIDADNFANRRMHHLKINREGRELTIQMDQQLRLSYNFSPEVEFRVIRSLTLGKVTENLGLDSEVAKANAMG
Specifications

Specifications

NP_570129
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DFVVVLLCKNGSLQVRYHLNKEETHVFTIDADNFANRRMHHLKINREGRELTIQMDQQLRLSYNFSPEVEFRVIRSLTLGKVTENLGLDSEVAKANAMG
RUO
CNTNAP5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
129684
CNTNAP5 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ31966/caspr5
CNTNAP5
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers