Learn More
Abnova™ Human CMTM4 Full-length ORF (NP_848933.1, 1 a.a. - 234 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00146223-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MRSGEELDGFEGEASSTSMISGASSPYQPTTEPVSQRRGLAGLRCDPDYLRGALGRLKVAQVILALIAFICIETIMACSPCEGLYFFEFVSCSAFVVTGVLLIMFSLNLHMRIPQINWNLTDLVNTGLSAFLFFIASIVLAALNHRAGAEIAAVIFGFLATAAYAVNTFLAVQKWRVSVRQQSTNDYIRARTESRDVDSRPEIQRLDTFSYSTNVTVRKKSPTNLLSLNHWQLASpecifications
NP_848933.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKLFSF4 | |
CMTM4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
146223 | |
CMTM4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRSGEELDGFEGEASSTSMISGASSPYQPTTEPVSQRRGLAGLRCDPDYLRGALGRLKVAQVILALIAFICIETIMACSPCEGLYFFEFVSCSAFVVTGVLLIMFSLNLHMRIPQINWNLTDLVNTGLSAFLFFIASIVLAALNHRAGAEIAAVIFGFLATAAYAVNTFLAVQKWRVSVRQQSTNDYIRARTESRDVDSRPEIQRLDTFSYSTNVTVRKKSPTNLLSLNHWQLA | |
RUO | |
CMTM4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |