missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CLU Partial ORF (NP_001822, 402 a.a. - 501 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001191-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: WKMLNTSSLLEQLNEQFNWVSRLANLTQGEDQYYLRVTTVASHTSDSDVPSGVTEVVVKLFDSDPITVTVPVEVSRKNPKFMETVAEKALQEYRKKHREESpecifications
NP_001822 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
WKMLNTSSLLEQLNEQFNWVSRLANLTQGEDQYYLRVTTVASHTSDSDVPSGVTEVVVKLFDSDPITVTVPVEVSRKNPKFMETVAEKALQEYRKKHREE | |
RUO | |
CLU | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1191 | |
CLU (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2 | |
CLU | |
Recombinant | |
wheat germ expression system |