missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Claudin 1 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0868
Additional Details : Weight : 0.01000kg
Description
CLDN1 Synthetic Peptide, PEP-0868, from Invitrogen.
This peptide corresponds to 20 amino acids near the carboxy terminus of human CLDN1.PEP-0868 can be used as a blocking peptide with polyclonal antibody PA5-20754.Specifications
9076 | |
purified | |
Human Claudin 1 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O95832 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
CLDN1 | |
0.02% sodium azide | |
50 μg | |
PBS |