missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHX10 Full-length ORF (NP_878314.1, 1 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00338917-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MTGKAGEALSKPKSETVAKSTSGGAPARCTGFGIQEILGLNKEPPSSHPRAALDGLAPGHLLAARSVLSPAGVGGMGLLGPGGLPGFYTQPTFLEVLSDPQSVHLQPLGRASGPLDTSQTASSDSEDVSSSDRKMSKSALNQTKKRKKRRHRTIFTSYQLEELEKAFNEAHYPDVYAREMLAMKTELPEDRIQVWFQNRRAKWRKREKCWGRSSVMAEYGLYGAMVRHSIPLPESILKSAKDGIMDSCAPWLLGMHKKSLEAAAESGRKPEGERQALPKLDKMEQDERGPDAQAAISQEELRENSIAVLRAKAQEHSTKVLGTVSGPDSLARSTEKPEEEEAMDEDRPAERLSPPQLEDMASpecifications
NP_878314.1 | |
Liquid | |
338917 | |
CHX10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTGKAGEALSKPKSETVAKSTSGGAPARCTGFGIQEILGLNKEPPSSHPRAALDGLAPGHLLAARSVLSPAGVGGMGLLGPGGLPGFYTQPTFLEVLSDPQSVHLQPLGRASGPLDTSQTASSDSEDVSSSDRKMSKSALNQTKKRKKRRHRTIFTSYQLEELEKAFNEAHYPDVYAREMLAMKTELPEDRIQVWFQNRRAKWRKREKCWGRSSVMAEYGLYGAMVRHSIPLPESILKSAKDGIMDSCAPWLLGMHKKSLEAAAESGRKPEGERQALPKLDKMEQDERGPDAQAAISQEELRENSIAVLRAKAQEHSTKVLGTVSGPDSLARSTEKPEEEEAMDEDRPAERLSPPQLEDMA | |
RUO | |
VSX2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHX10/HOX10/MCOP2/MCOPCB3/RET1 | |
VSX2 | |
Yes | |
wheat germ expression system |