Learn More
Abnova™ Human CHKB Full-length ORF (NP_005189.2, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001120-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Choline kinase (CK) and ethanolamine kinase (EK) catalyze the phosphorylation of choline/ethanolamine to phosphocholine/phosphoethanolamine. This is the first enzyme in the biosynthesis of phosphatidylcholine/phosphatidylethanolamine in all animal cells. The highly purified CKs from mammalian sources and their recombinant gene products have been shown to have EK activity also, indicating that both activities reside on the same protein. The choline kinase-like protein encoded by CHKL belongs to the choline/ethanolamine kinase family; however, its exact function is not known. Read-through transcripts are expressed from this locus that include exons from the downstream CPT1B locus. [provided by RefSeq]
Sequence: MAAEATAVAGSGAVGGCLAKDGLQQSKCPDTTPKRRRASSLSRDAERRAYQWCREYLGGAWRRVQPEELRVYPVSGGLSNLLFRCSLPDHLPSVGEEPREVLLRLYGAILQGVDSLVLESVMFAILAERSLGPQLYGVFPEGRLEQYIPSRPLKTQELREPVLSAAIATKMAQFHGMEMPFTKEPHWLFGTMERYLKQIQDLPPTGLPEMNLLEMYSLKDEMGNLRKLLESTPSPVVFCHNDIQEGNILLLSEPENADSLMLVDFEYSSYNYRGFDIGNHFCEWVYDYTHEEWPFYKARPTDYPTQEQQLHFIRHYLAEAKKGETLSQEEQRKLEEDLLVEVSRYALASHFFWGLWSILQASMSTIEFGYLDYAQSRFQFYFQQKGQLTSVHSSSSpecifications
NP_005189.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHETK/CHKL/CKEKB/EKB | |
CHKB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1120 | |
CHKB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAEATAVAGSGAVGGCLAKDGLQQSKCPDTTPKRRRASSLSRDAERRAYQWCREYLGGAWRRVQPEELRVYPVSGGLSNLLFRCSLPDHLPSVGEEPREVLLRLYGAILQGVDSLVLESVMFAILAERSLGPQLYGVFPEGRLEQYIPSRPLKTQELREPVLSAAIATKMAQFHGMEMPFTKEPHWLFGTMERYLKQIQDLPPTGLPEMNLLEMYSLKDEMGNLRKLLESTPSPVVFCHNDIQEGNILLLSEPENADSLMLVDFEYSSYNYRGFDIGNHFCEWVYDYTHEEWPFYKARPTDYPTQEQQLHFIRHYLAEAKKGETLSQEEQRKLEEDLLVEVSRYALASHFFWGLWSILQASMSTIEFGYLDYAQSRFQFYFQQKGQLTSVHSSS | |
RUO | |
CHKB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |