Learn More
Abnova™ Human CEP2 Partial ORF (AAH01433, 141 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011190-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a core centrosomal protein required for centriole-centriole cohesion during interphase of the cell cycle. The encoded protein dissociates from the centrosomes when parental centrioles separate at the beginning of mitosis. The protein associates with and is phosphorylated by NIMA-related kinase 2, which is also associated with the centrosome. [provided by RefSeq]
Sequence: LTVDWSRARDELMRKESQWQMEQEFFKGYLKGEHGRLLSLWREVVTFRRHFLEMKSATDRDLMELKAEHVRLSGSLLTCCLRLTVGAQSREPNGSGRMDGSpecifications
AAH01433 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LTVDWSRARDELMRKESQWQMEQEFFKGYLKGEHGRLLSLWREVVTFRRHFLEMKSATDRDLMELKAEHVRLSGSLLTCCLRLTVGAQSREPNGSGRMDG | |
RUO | |
CEP250 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11190 | |
CEP2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C-NAP1/CEP2/CNAP1/MGC88542 | |
CEP250 | |
Recombinant | |
wheat germ expression system |