Learn More
Abnova™ Human CENTG2 Partial ORF (NP_055729.2, 672 a.a. - 781 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00116987-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
CENTG2 belongs to an ADP-ribosylation factor GTPase-activating (ARF-GAP) protein family involved in membrane traffic and actin cytoskeleton dynamics (Nie et al., 2002 [PubMed 12388557]).[supplied by OMIM]
Sequence: LPCTELSLGQHLLRATADEDLRTAILLLAHGSRDEVNETCGEGDGRTALHLACRKGNVVLAQLLIWYGVDVTARDAHGNTALAYARQASSQECIDVLLQYGCPDERFVLMSpecifications
NP_055729.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPCTELSLGQHLLRATADEDLRTAILLLAHGSRDEVNETCGEGDGRTALHLACRKGNVVLAQLLIWYGVDVTARDAHGNTALAYARQASSQECIDVLLQYGCPDERFVLM | |
RUO | |
AGAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
116987 | |
CENTG2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CENTG2/GGAP1/KIAA1099/MGC71657 | |
AGAP1 | |
Recombinant | |
wheat germ expression system |