Learn More
Abnova™ Human CDON Partial ORF (NP_058648, 1155 a.a. - 1263 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00050937-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
CDON and BOC (MIM 608708) are cell surface receptors of the immunoglobulin (Ig)/fibronectin type III (FNIII; see MIM 135600) repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.[supplied by OMIM]
Sequence: VKVPVCLTSAVPDCGQLPEESVKDNVEPVPTQRTCCQDIVNDVSSDGSEDPAEFSRGDSCAHSETEINIVSWNALILPPVPEGCAEKTMWSPPGIPLDSPTEVLQQPRESpecifications
NP_058648 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VKVPVCLTSAVPDCGQLPEESVKDNVEPVPTQRTCCQDIVNDVSSDGSEDPAEFSRGDSCAHSETEINIVSWNALILPPVPEGCAEKTMWSPPGIPLDSPTEVLQQPRE | |
RUO | |
CDON | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50937 | |
CDON (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDO/MGC111524/ORCAM | |
CDON | |
Recombinant | |
wheat germ expression system |