missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDCA7L Full-length ORF (NP_061189.2, 1 a.a. - 454 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055536-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141533

  • 6270.00 NOK / 25µg
Estimated Shipment: 11-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MELATRYQIPKEVADIFNAPSDDEEFVGFRDDVPMETLSSEESCDSFDSLESGKQQDVRFHSKYFTEELRRIFIEDTDSETEDFAGFTQSDLNGKTNPEVMVVESDLSDDGKASLVSEEEEDEEEDKATPRRSRSRRSSIGLRVAFQFPTKKLANKPDKNSSSEQLFSSARLQNEKKTILERKKDCRQVIQREDSTSESEDDSRDESQESSDALLKRTMNIKENKAMLAQLLAELNSMPDFFPVRTPTSASRKKTVRRAFSEGQITRRMNPTRSARPPEKFALENFTVSAAKFAEEFYSFRRRKTIGGKCREYRRRHRISSFRPVEDITEEDLENVAITVRDKIYDKVLGNTCHQCRQKTIDTKTVCRNQGCCGVRGQFCGPCLRNRYGEDVRSALLDPDWVCPPCRGICNCSYCRKRDGRCATGILIHLAKFYGYDNVKEYLESLQKELVEDN
Specifications

Specifications

NP_061189.2
Liquid
55536
CDCA7L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MELATRYQIPKEVADIFNAPSDDEEFVGFRDDVPMETLSSEESCDSFDSLESGKQQDVRFHSKYFTEELRRIFIEDTDSETEDFAGFTQSDLNGKTNPEVMVVESDLSDDGKASLVSEEEEDEEEDKATPRRSRSRRSSIGLRVAFQFPTKKLANKPDKNSSSEQLFSSARLQNEKKTILERKKDCRQVIQREDSTSESEDDSRDESQESSDALLKRTMNIKENKAMLAQLLAELNSMPDFFPVRTPTSASRKKTVRRAFSEGQITRRMNPTRSARPPEKFALENFTVSAAKFAEEFYSFRRRKTIGGKCREYRRRHRISSFRPVEDITEEDLENVAITVRDKIYDKVLGNTCHQCRQKTIDTKTVCRNQGCCGVRGQFCGPCLRNRYGEDVRSALLDPDWVCPPCRGICNCSYCRKRDGRCATGILIHLAKFYGYDNVKEYLESLQKELVEDN
RUO
CDCA7L
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp762L0311/JPO2/R1/RAM2
CDCA7L
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers