missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CD96 His-tag, Recombinant Protein

HEK293 cells Recombinant Protein

Brand:  Invitrogen™ A42596

Additional Details : Weight : 0.01070kg

 View more versions of this product

Product Code. 17144263

  • 2890.00 NOK / 50µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD96 is a cell adhesion receptor also known as TACTILE. A member of the Ig superfamily, CD96 is a single pass transmembrane protein consisting of three extracellular Ig-like domains and a proline/serine/threonine rich region. CD96 is expressed on resident monocytes and T cells at all developmental stages (including gamma-delta T cells, thymocytes, and NK cells). In the mouse, this receptor is not expressed on B cells. Unlike in humans, mouse CD96 does not exist as alternative splice variants. Reports indicate that CD96 binds CD155/poliovirus receptor and Nectin-1 to mediate NK cell adhesion to target cells and cytotoxicity.
Specifications

Specifications

P40200
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Human CD96 His-tag
HEK293 cells
RUO
1700109I12Rik; CD96; CD96 antigen; CD96 molecule; cell surface antigen CD96; DKFZp667E2122; sCD96; soluble CD96; T cell activation, increased late expression; t cell-activated increased late expression protein; TACTILE; T-cell surface protein tactile
Human
Recombinant
≥ 85%
Control, ELISA, Immunohistochemistry, Western Blot
10225
50 μg
-20°C
<1 EU/μg
Cd96
Unconjugated
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers