missing translation for 'onlineSavingsMsg'
Learn More

Human CD90/Thy1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

1885.00 NOK - 4705.00 NOK

Specifications

Antigen CD90/Thy1
Clone 783922
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18750107
GREENER_CHOICE
View Documents
R&D Systems
MAB20671-SP
25 μg
1885.00 NOK
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18772437
GREENER_CHOICE
View Documents
R&D Systems
MAB20671-100
100 μg
4705.00 NOK
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD90/Thy1 Monoclonal specifically detects CD90/Thy1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

CD90/Thy1
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Monoclonal
Supernatant
Human
P04216
7070
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
783922
Flow Cytometry, CyTOF
Unconjugated
Mouse
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Chinese Hamster Ovary cell line, CHO-derived human CD90/THY1, Met1-Ser141, Accession # P04216
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CD90/THY1 in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.