Learn More
Abnova™ Human CD300E Full-length ORF (AAI00889.1, 1 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00342510-P01.25ug
Additional Details : Weight : 0.00010kg
Description
CD300LE is an activating receptor of the immunoglobulin (Ig) superfamily expressed on myeloid cells. It mediates activating signals by interacting with DAP12 (TYROBP; MIM 604142) (Aguilar et al., 2004 [PubMed 15557162]).[supplied by OMIM]
Sequence: MWLLPALLLLCLSGCLSLKGPGSVTGTAGDSLTVWCQYESMYKGYNKYWCRGQYDTSCESIVETKGEEKVERNGRVSIRDHPEALAFTVTMQNLNEDDAGSYWCKIQTVWVLDSWSRDPSDLVRVYVSPAITTPRRTTHPATPPIFLVVNPGRNLSTREVLTQNSGFRLSSPHFLLVVLLKLPLLLSMLGAVFWVNRPQWAPPGRSpecifications
AAI00889.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD300LE/CLM2/IREM2 | |
CD300E | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
342510 | |
CD300E (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWLLPALLLLCLSGCLSLKGPGSVTGTAGDSLTVWCQYESMYKGYNKYWCRGQYDTSCESIVETKGEEKVERNGRVSIRDHPEALAFTVTMQNLNEDDAGSYWCKIQTVWVLDSWSRDPSDLVRVYVSPAITTPRRTTHPATPPIFLVVNPGRNLSTREVLTQNSGFRLSSPHFLLVVLLKLPLLLSMLGAVFWVNRPQWAPPGR | |
RUO | |
CD300E | |
Wheat Germ (in vitro) | |
GST | |
Liquid |