missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CCT2 Partial ORF (NP_006422, 117 a.a. - 217 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010576-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16145362

  • 6245.00 NOK / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the gene described in this record have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: IAKKIHPQTIIAGWREATKAAREALLSSAVDHGSDEVKFRQDLMNIAGTTLSSKLLTHHKDHFTKLAVEAVLRLKGSGNLEAIHIIKKLGGSLADSYLDEG
Specifications

Specifications

NP_006422
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.85kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
IAKKIHPQTIIAGWREATKAAREALLSSAVDHGSDEVKFRQDLMNIAGTTLSSKLLTHHKDHFTKLAVEAVLRLKGSGNLEAIHIIKKLGGSLADSYLDEG
RUO
CCT2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10576
CCT2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
99D8.1/CCT-beta/CCTB/MGC142074/MGC142076/PRO1633/TCP-1-beta
CCT2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers