missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR2 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB151G-025
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Flow Cytometry.
Specifications
| CCR2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NP_001116868 | |
| CCR2 | |
| NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 | |
| 25 Tests | |
| 729230 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 48607 | |
| Flow Cytometry 5 uL/10^6 cells | |
| C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human CCR-2 transfectants but not the parental cell line or CCR-5 transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction