missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL3 (MIP-1 alpha) Recombinant Protein
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
Description
Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.
Specifications
Specifications
| Accession Number | P10147 |
| Concentration | 200 μg/mL |
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | 4 mM HCl with no preservative |
| Gene ID (Entrez) | 6348 |
| Molecular Weight (g/mol) | 7.5 kD |
| Name | Human MIP-1 alpha (aa 27-92) |
| Quantity | 500 μg |
| Source | E. coli |
| Storage Requirements | -20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction