missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCL3 (MIP-1 alpha) Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87850
Additional Details : Weight : 0.01000kg
Description
Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.Specifications
P10147 | |
Control, ELISA, Western Blot | |
6348 | |
Human MIP-1 alpha (aa 27-92) | |
E. coli | |
RUO | |
AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78alpha; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1 alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); Mip1a; MIP-1 A; MIP-1 alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; SCYA3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1 A); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5 | |
Human | |
Recombinant | |
>95% |
200 μg/mL | |
4 mM HCl with no preservative | |
7.5 kD | |
25 μg | |
-20°C | |
0.01 ng/ μg cytokine | |
Ccl3 | |
Unconjugated | |
Liquid |