missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCL3 (MIP-1 alpha) Recombinant Protein

Product Code. 17636645
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Unit Size:
100µg
10µg
25µg
500µg
This item is not returnable. View return policy

Product Code. 17636645

Brand: Invitrogen™ RP87849

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

Macrophage inflammatory protein-1a (MIP-1a) is an 8kDa member of the C-C chemokine subfamily. C-C chemokines are chemoattractants and activators for monocytes and T cells. MIP-1a has been identified as one of the major HIV-SFs produced by CD8+ T cells, along with MIP-1b and RANTES.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P10147
Concentration 200 μg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation 4 mM HCl with no preservative
Gene ID (Entrez) 6348
Molecular Weight (g/mol) 7.5 kD
Name Human MIP-1 alpha (aa 27-92)
Quantity 10 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration 0.01 ng/ μg cytokine
Gene Alias AI323804; C-C motif chemokine 3; C-C motif chemokine ligand 3; CCL3; chemokine (C-C motif) ligand 3; G0/G1 switch regulatory protein 19-1; G0S19-1; Heparin-binding chemotaxis protein; H-MIP-1-alpha; L2G25B; LD78alpha; LD78-alpha; LD78-alpha(4-69); Macrophage inflammatory protein 1 alpha (Small inducible cytokine A3); macrophage inflammatory protein 1-alpha; macrophage inflammatory protein-1 alpha; MIP1 (a); mip1 alpha; MIP-1 alpha; MIP1-(a); Mip1a; MIP-1 A; MIP-1 alpha; MIP-1-alpha; MIP1-alpha; MIP-1-alpha(4-69); PAT 464.1; RP23-320E6.7; SCYA3; SIS-alpha; SIS-beta; small inducible cytokine A3; small inducible cytokine A3 (homologous to mouse Mip-1 A); small-inducible cytokine A3; Tonsillar lymphocyte LD78 alpha protein; TY-5
Gene Symbol Ccl3
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Liquid
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.