Learn More
Human CCL3/CCL4 Antibody, R&D Systems™
GREENER_CHOICE
Product Code. 18742597
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
Product Code. Quantity unitSize
18742597 25 μg 25µg
18724368 100 μg 100µg
18784637 500 μg 500µg
3 options
Product Code. 18742597
Brand: R&D Systems MAB270SP
Mouse Monoclonal Antibody has been used in 8 publications
CCL3/CCL4 Monoclonal specifically detects CCL3/CCL4 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Antigen CCL3/CCL4
Applications Western Blot, Neutralization, Immunocytochemistry
Classification Monoclonal
Clone 93321
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.075-0.375 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10147
Gene Symbols CCL3
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL3/MIP-1 alpha Ala27-Ala92 Accession # P10147
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects human CCL3/MIP-1 alpha in direct ELISAs and Western blots. In direct ELISAs, greater than 75% cross-reactivity with recombinant human (rh) CCL4/MIP-1 beta is observed while approximately 30% cross-reactivity with recombinant mouse (rm) CCL3, rmCCL4/MIP-1 beta , and rhCCL14 is observed. No cross-reactivity with rhCCL1, 2, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, rmCCL1, 2, 5, 6, 7, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, or recombinant rat CCL20 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1